Methyl-5-fluoro-2-nitrobenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61307-14
H61307-06
CAAAH61307-14EA
368.14
CAD
CAAAH61307-14
AAH61307-06
Methyl-5-fluoro-2-nitrobenzoate 98%
Methyl 5-fluoro-2-nitrobenzoate
Methyl 5-fluoro-2-nitrobenzoate, 98%
Formula:
C₈H₆FNO₄ Melting Pt: 35…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD03425619 CAS Number: 393-85-1 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...