5-Methylpyrazin-2-amine 98%

Supplier: Thermo Scientific Chemicals
Warning

H61286-03 H61286-MD
CAAAH61286-03EA 146.47 CAD
CAAAH61286-03 CAAAH61286-MD
5-Methylpyrazin-2-amine 98%
5-Methylpyrazin-2-amine

2-Amino-5-methylpyrazine, 98%


Purity: 98% CAS No.: 5521-58-4 Molecular Formula: C5H7N3 Formula Weight: 109.13 Physical Form: Solid MDL No.: MFCD08437664

Formula: C5H7N3
Storage Temperature: Ambient
MDL Number: MFCD08437664
CAS Number: 5521-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR