5'-Bromo-2'-fluoroacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(5-Bromo-2-fluorophenyl)ethanone

H61282-06 H61282-14
CAAAH61282-06EA 141.57 CAD
CAAAH61282-06 CAAAH61282-14
5'-Bromo-2'-fluoroacetophenone 98%
5'-Bromo-2'-fluoroacetophenone

5'-Bromo-2'-fluoroacetophenone, 98%

Formula: C₈H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD11226880
CAS Number: 198477-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR