Ethyl-2-aminoisonicotinate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-isonicotinic acid ethyl ester, Ethyl 2-aminopyridine-4-carboxylate, 2-Aminopyridine-4-carboxylic acid Ethyl Ester, Ethyl 2-aminoisonicotinate, Ethyl 2-Aminopyridine-4-carboxylate, 2-Aminoisonicotinic acid Ethyl Ester, Ethyl 2-Aminoisonicotinate

H61280-06 H61280-14
CAAAH61280-06EA 152.79 CAD
CAAAH61280-06 CAAAH61280-14
Ethyl-2-aminoisonicotinate 97%
Ethyl 2-aminoisonicotinate

Ethyl 2-aminopyridine-4-carboxylate, 97%

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03791260
CAS Number: 13362-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR