2-Fluoro-3-nitropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

H61260-14 H61260-22
CAAAH61260-14EA 150.1 CAD
CAAAH61260-14 CAAAH61260-22
2-Fluoro-3-nitropyridine 98%
2-Fluoro-3-nitropyridine

2-Fluoro-3-nitropyridine, 98%

Formula: C₅H₃FN₂O₂
Boiling Pt: 110 °C/10 mm
Melting Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD03095068
CAS Number: 1480-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR