2-Bromo-6-fluorobenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61251-03
H61251-06
CAAAH61251-03EA
102.54
CAD
CAAAH61251-03
CAAAH61251-06
2-Bromo-6-fluorobenzoic acid 97%
2-Bromo-6-fluorobenzoic acid
2-Bromo-6-fluorobenzoic acid, 97%
Formula:
C₇H₄BrFO₂ Melting Pt: 154…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD01569539 CAS Number: 2252-37-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...