3,6-Dibromopyridazine 95%

Supplier: Thermo Scientific Chemicals
Warning

H61223-03 H61223-06
CAAAH61223-03EA 70.63 CAD
CAAAH61223-03 CAAAH61223-06
3,6-Dibromopyridazine 95%
3,6-Dibromopyridazine

3,6-Dibromopyridazine, 95%

Formula: C₄H₂Br₂N₂
Melting Pt: 117…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00233947
CAS Number: 17973-86-3
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR