2-Morpholinoacetic acid hydrochloride 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61188-03
H61188-MD
H61188-06
CAAAH61188-03EA
102.07
CAD
CAAAH61188-03
CAAAH61188-MD
AAH61188-06
2-Morpholinoacetic acid hydrochloride 95%
2-Morpholinoacetic acid hydrochloride
4-Morpholineacetic acid hydrochloride, 95%
Purity: 95%
CAS No.: 89531-58-8
Molecular Formula: C6H11NO3.HCl
Formula Weight: 181.62
Physical Form: Powder
Appearance: White
Beilstein Reference: 3701710
MDL No.: MFCD03540888
Formula:
C6H11NO3 Storage Temperature: Ambient |
MDL Number:
MFCD03540888 CAS Number: 3235-69-6 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...