4-Bromo-2-fluorophenylacetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61176-06
H61176-14
CAAAH61176-06EA
192.76
CAD
CAAAH61176-06
CAAAH61176-14
4-Bromo-2-fluorophenylacetic acid 98%
4-Bromo-2-fluorophenylacetic acid
4-Bromo-2-fluorophenylacetic acid, 98%
Formula:
C₈H₆BrFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD09032952 CAS Number: 114897-92-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...