2-Bromo-4-nitroimidazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61155-03
H61155-06
CAAAH61155-03EA
57.99
CAD
CAAAH61155-03
CAAAH61155-06
CAAAH61155-14
2-Bromo-4-nitroimidazole 98%
2-Bromo-4-nitroimidazole
2-Bromo-5-nitroimidazole, 98%
Formula:
C₃H₂BrN₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD09025308 CAS Number: 65902-59-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...