2-Chloro-5-nitroimidazole 95%

Supplier: Thermo Scientific Chemicals
Warning

H61141-03 H61141-06
CAAAH61141-03EA 118.82 CAD
CAAAH61141-03 CAAAH61141-06 CAAAH61141-MD
2-Chloro-5-nitroimidazole 95%
2-Chloro-5-nitroimidazole

2-Chloro-5-nitroimidazole, 95%


Purity: 95% CAS No.: 57531-37-0 Molecular Formula: C3H2ClN3O2 Formula Weight: 147.52 Physical Form: Solid MDL No.: MFCD08705680

Formula: C₃H₂ClN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD03419295
CAS Number: 57531-37-0
EINECS: 611-554-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR