4'-Bromo-2'-hydroxyacetophenone 95%

Supplier: Thermo Scientific Chemicals
Warning

H61137-03 H61137-06
CAAAH61137-03EA 135.25 CAD
CAAAH61137-03 CAAAH61137-06 CAAAH61137-MD
4'-Bromo-2'-hydroxyacetophenone 95%
4'-Bromo-2'-hydroxyacetophenone

4'-Bromo-2'-hydroxyacetophenone, 95%

Formula: C₈H₇BrO₂
Storage Temperature: Ambient
MDL Number: MFCD03428533
CAS Number: 30186-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR