Methyl-4-amino-3-methylbenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 4-amino-3-methylbenzoate

H61117-14 H61117-22
CAAAH61117-14EA 85.64 CAD
CAAAH61117-14 CAAAH61117-22
Methyl-4-amino-3-methylbenzoate 98%
Methyl 4-amino-3-methylbenzoate

Methyl 4-amino-3-methylbenzoate, 98%


Purity: 98% CAS No.: 18595-14-7 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Physical Form: Solid Melting Point: 116-120° MDL No.: MFCD00102230

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 116…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00102230
CAS Number: 18595-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR