4-Fluoro-2-nitroanisole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Fluoro-1-methoxy-2-nitrobenzene

H61110-06 H61110-14
CAAAH61110-06EA 74.42 CAD
CAAAH61110-06 CAAAH61110-14
4-Fluoro-2-nitroanisole 98%
4-Fluoro-2-nitroanisole

4-Fluoro-2-nitroanisole, 98%

Formula: C₇H₆FNO₃
Melting Pt: 62…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00013375
CAS Number: 445-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR