Triphenylantimony dichloride 99%

Supplier: Thermo Scientific Chemicals
Danger

H55719-06 H55719-14
CAAAH55719-06EA 83.42 CAD
CAAAH55719-06 CAAAH55719-14
Triphenylantimony dichloride 99%
Triphenylantimony dichloride

Triphenylantimony(V) dichloride, 99%

Formula: C₁₈H₁₅Cl₂Sb
Melting Pt: 143…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00013598
CAS Number: 594-31-0
EINECS: 209-834-1
UN: 3467
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR