2,2'-Bithiophene-5-boronic acid pinacol ester 98%
Supplier: Thermo Scientific Chemicals
H37989-03
H37989-06
CAAAH37989-03EA
73.63
CAD
CAAAH37989-03
CAAAH37989-06
2,2'-Bithiophene-5-boronic acid pinacol ester 98%
2,2'-Bithiophene-5-boronic acid pinacol ester
2,2'-Bithiophene-5-boronic acid pinacol ester, 98%
Formula:
C₁₄H₁₇BO₂S₂ Melting Pt: 35…38 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04039974 CAS Number: 479719-88-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...