6-Bromo-3,4-dihydro-1H-quinolin-2-one 98%

Supplier: Thermo Scientific Chemicals
Warning

H37968-03 H37968-06 H37968-14
CAAAH37968-03EA 33.81 CAD
CAAAH37968-03 CAAAH37968-06 CAAAH37968-14
6-Bromo-3,4-dihydro-1H-quinolin-2-one 98%
6-Bromo-3,4-dihydro-1H-quinolin-2-one

6-Bromo-3,4-dihydro-2(1H)-quinolinone, 98%


Purity: 98% CAS No.: 3279-90-1 Molecular Formula: C9H8BrNO Formula Weight: 226.07 Physical Form: Powder Sensitivity: Light Sensitive Appearance: Brown Melting Point: 170-172° MDL No.: MFCD03839832

Formula: C₉H₈BrNO
MW: 226.07 g/mol
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD03839832
CAS Number: 3279-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR