Sand, washed

Supplier: Thermo Scientific Chemicals

H37933-0B H37933-0I
CAAAH37933-0BEA 80.9 CAD
CAAAH37933-0B CAAAH37933-0I
Sand, washed
Sand

Sand, washed


CAS No.: 14808-60-7 Molecular Formula: O2Si Formula Weight: 60.08 Physical Form: Granules Appearance: Pale cream/pale brown Odor: Odorless Melting Point: 1710° Boiling Point: 2230° MDL No.: MFCD02100519

Formula: SiO₂
Boiling Pt: 2230 °C
Melting Pt: 1710 °C
Storage Temperature: Ambient
MDL Number: MFCD02100519
CAS Number: 7631-86-9
EINECS: 238-878-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR