2,2':6',2''-Terpyridine-4'-carboxylic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

H37760-03 H37760-MD
CAAAH37760-03EA 398.16 CAD
CAAAH37760-03 CAAAH37760-MD
2,2':6',2''-Terpyridine-4'-carboxylic acid 95%
2,2':6',2''-Terpyridine-4'-carboxylic acid

2,2':6',2''-Terpyridine-4'-carboxylic acid, 95%


Purity: 95% CAS No.: 148332-36-9 Molecular Formula: C16H11N3O2 Formula Weight: 277.28 Physical Form: Powder Appearance: White to cream Melting Point: 280-282° MDL No.: MFCD04114290

Formula: C₁₆H₁₁N₃O₂
Melting Pt: 280…282 °C
Storage Temperature: Ambient
MDL Number: MFCD04114290
CAS Number: 148332-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR