1,3,5-Tris(4-bromophenyl)benzene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H37636-03
H37636-06
CAAAH37636-03EA
95.75
CAD
CAAAH37636-03
CAAAH37636-06
1,3,5-Tris(4-bromophenyl)benzene 97%
1,3,5-Tris(4-bromophenyl)benzene
1,3,5-Tris(4-bromophenyl)benzene, 97%
Purity: 97%
CAS No.: 7511-49-1
Molecular Formula: C24H15Br3
Formula Weight: 543.10
Physical Form: Crystalline powder
Appearance: Yellow to pale brown
Melting Point: 261-265°
MDL No.: MFCD00362911
Formula:
C₂₄H₁₅Br₃ Melting Pt: 261…265 °C Storage Temperature: Ambient |
MDL Number:
MFCD00362911 CAS Number: 7511-49-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...