Ethyl-2-acetylisonicotinate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H36911-03
H36911-06
CAAAH36911-03EA
146.31
CAD
CAAAH36911-03
CAAAH36911-06
Ethyl-2-acetylisonicotinate 96%
Ethyl-2-acetylisonicotinate
Ethyl 2-acetylisonicotinate, 96%
Purity: 96%
CAS No.: 25028-32-4
Molecular Formula: C10H11NO3
Formula Weight: 193.20
Physical Form: Fused solid
Appearance: Cream
Melting Point: 52-55°
MDL No.: MFCD11042531
Formula:
C₁₀H₁₁NO₃ Melting Pt: 52…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD11042531 CAS Number: 25028-32-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...