5-Methoxyresorcinol 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61996-03
H61996-06
H61996-MD
CAAAH61996-03EA
122.77
CAD
CAAAH61996-03
CAAAH61996-06
CAAAH61996-MD
5-Methoxyresorcinol 95%
5-Methoxyresorcinol
5-Methoxyresorcinol, 95%
Purity: 95%
CAS No.: 2174-64-3
Molecular Formula: C7H8O3
Formula Weight: 140.14
Physical Form: Solid
Appearance: Yellow
Melting Point: 78-80°
Boiling Point: 188-189°/12mm
Beilstein Reference: 1423578
MDL No.: MFCD00002285
Formula:
C₇H₈O₃ Boiling Pt: 188…189 °C/12 mm Melting Pt: 78…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002285 CAS Number: 2174-64-3 EINECS: 218-532-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...