5'-Bromo-2'-hydroxyacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61979-09
H61979-18
CAAAH61979-09EA
82.95
CAD
CAAAH61979-09
CAAAH61979-18
5'-Bromo-2'-hydroxyacetophenone 98%
5'-Bromo-2'-hydroxyacetophenone
5'-Bromo-2'-hydroxyacetophenone, 98%
Formula:
C₈H₇BrO₂ Melting Pt: 56…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191850 CAS Number: 1450-75-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...