4-Bromo-3-chlorobenzotrifluoride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-2-chloro-4-trifluoromethylbenzene
H61978-06
H61978-14
CAAAH61978-06EA
65.89
CAD
CAAAH61978-06
CAAAH61978-14
CAAAH61978-22
4-Bromo-3-chlorobenzotrifluoride 98%
4-Bromo-3-chlorobenzotrifluoride
4-Bromo-3-chlorobenzotrifluoride, 98%
Formula:
C₇H₃BrClF₃ MW: 259.45 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00673985 CAS Number: 402-04-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...