4-Bromo-3-nitrobenzoic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

H61975-06 H61975-14
CAAAH61975-06EA 91.32 CAD
CAAAH61975-06 CAAAH61975-14
4-Bromo-3-nitrobenzoic acid 95%
4-Bromo-3-nitrobenzoic acid

4-Bromo-3-nitrobenzoic acid, 95%

Formula: C₇H₄BrNO₄
Melting Pt: 199…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00272137
CAS Number: 6319-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR