Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0) Pd 12-14%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61964-03
H61964-06
CAAAH61964-03EA
147.57
CAD
CAAAH61964-03
CAAAH61964-06
Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0) Pd 12-14%
Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0)
Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0), Pd 13%
CAS No.: 811862-77-8
Molecular Formula: C42H44O10Pd
Formula Weight: 815.23
Physical Form: Powder
Appearance: Dark red
Melting Point: 170-178°
MDL No.: MFCD07369799
Formula:
C₄₂H₄₄O₁₀Pd MW: 815.224 g/mol Melting Pt: 170…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD07369799 CAS Number: 811862-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...