2,5-Dibromo-3-nitropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

H61961-14 H61961-22
CAAAH61961-06EA 64.89 CAD
CAAAH61961-06 CAAAH61961-14 CAAAH61961-22
2,5-Dibromo-3-nitropyridine 98%
2,5-Dibromo-3-nitropyridine

2,5-Dibromo-3-nitropyridine, 98%

Formula: C₅H₂Br₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09266223
CAS Number: 15862-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR