2-Amino-6-chloropyrazine 95%

Supplier: Thermo Scientific Chemicals
Warning

H61934-14 H61934-22
CAAAH61934-06EA 90.63 CAD
CAAAH61934-06 CAAAH61934-14 CAAAH61934-22
2-Amino-6-chloropyrazine 95%
2-Amino-6-chloropyrazine

2-Amino-6-chloropyrazine, 95%

Formula: C₄H₄ClN₃
Storage Temperature: Ambient
MDL Number: MFCD00055024
CAS Number: 33332-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR