3,5-Dimethyl-4-isoxazolecarbaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61912-06
H61912-14
CAAAH61912-06EA
224.36
CAD
CAAAH61912-06
CAAAH61912-14
3,5-Dimethyl-4-isoxazolecarbaldehyde 97%
3,5-Dimethyl-4-isoxazolecarbaldehyde
3,5-Dimethylisoxazole-4-carboxaldehyde, 97%
Purity: 97%
CAS No.: 54593-26-9
Molecular Formula: C6H7NO2
Formula Weight: 125.13
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 35-38°
MDL No.: MFCD02681977
Formula:
C6H7NO2 Melting Pt: 35…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD02681977 CAS Number: 54593-26-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...