6-Chloro-1H-indole-3-carbaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Chloroindole-3-carboxaldehyde

H61911-06 H61911-14
CAAAH61911-06EA 122.13 CAD
CAAAH61911-06 CAAAH61911-14
6-Chloro-1H-indole-3-carbaldehyde 98%
6-Chloro-1H-indole-3-carbaldehyde

6-Chloroindole-3-carboxaldehyde, 98%


Purity: 98% CAS No.: 703-82-2 Molecular Formula: C9H6ClNO Formula Weight: 179.60 Physical Form: Powder Appearance: Pale brown MDL No.: MFCD06657152

Formula: C₉H₆ClNO
Storage Temperature: Ambient
MDL Number: MFCD06657152
CAS Number: 703-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR