Methyl-3-methyl-2-nitrobenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 3-methyl-2-nitrobenzoate
H61907-14
H61907-22
CAAAH61907-14EA
98.91
CAD
CAAAH61907-14
CAAAH61907-22
Methyl-3-methyl-2-nitrobenzoate 98%
Methyl 3-methyl-2-nitrobenzoate
Methyl 3-methyl-2-nitrobenzoate, 98%
Purity: 98%
CAS No.: 5471-82-9
Molecular Formula: C9H9NO4
Formula Weight: 195.17
Physical Form: Solid
MDL No.: MFCD01003947
Formula:
C₉H₉NO₄ MW: 195.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01003947 CAS Number: 5471-82-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...