3-Fluoro-2-formylpyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Fluoropyridine-2-carboxaldehyde, 3-Fluoropicolinaldehyde, 3-Fluoropyridine-2-carbaldehyde, 3-Fluoro-2-pyridinecarboxaldehyde

H61902-03 H61902-06
CAAAH61902-03EA 115.02 CAD
CAAAH61902-03 CAAAH61902-06 CAAAH61902-MD
3-Fluoro-2-formylpyridine 98%
3-Fluoro-2-formylpyridine

3-Fluoropyridine-2-carboxaldehyde, 98%

Formula: C₆H₄FNO
MW: 125.1
Storage Temperature: Ambient
MDL Number: MFCD07781234
CAS Number: 31224-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR