2',4'-Difluoro-2-(1H-1,2,4-triazol-1-yl)acetophenone 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(2,4-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone

H61887-06 H61887-14
CAAAH61887-06EA 101.59 CAD
CAAAH61887-06 CAAAH61887-14
2',4'-Difluoro-2-(1H-1,2,4-triazol-1-yl)acetophenone 97%
2',4'-Difluoro-2-(1H-1,2,4-triazol-1-yl)acetophenone

2',4'-Difluoro-2-(1H-1,2,4-triazol-1-yl)acetophenone, 97%

Formula: C₁₀H₇F₂N₃O
MW: 223.18 g/mol
Melting Pt: 103…107 °C
Storage Temperature: Ambient
MDL Number: MFCD02093825
CAS Number: 86404-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR