2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]benzimidazole, 2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)thio)-1H-benzo[d]imidazole, Lansoprazole sulphide, Lansoprazole sulfide
2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole, 98%
Purity: 98%
CAS No.: 103577-40-8
Molecular Formula: C16H14F3N3OS
Formula Weight: 353.36
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline powder
Appearance: Off-white
MDL No.: MFCD00834357
Formula:
C₁₆H₁₄F₃N₃OS MW: 353.363 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00834357 CAS Number: 103577-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...