Methyl-5-bromo-2-methylbenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 5-bromo-2-methylbenzoate, 5-Bromo-2-methylbenzoic acid methyll ester, 5-Bromo-o-toluic acid methyll ester, Methyl 5-Bromo-o-toluate, methyl 5-bromo-2-methyl-benzoate

H61861-06 H61861-14 H61861-22
CAAAH61861-06EA 96.06 CAD
CAAAH61861-06 CAAAH61861-14 CAAAH61861-22
Methyl-5-bromo-2-methylbenzoate 98%
Methyl-5-bromo-2-methylbenzoate

Methyl 5-bromo-2-methylbenzoate, 98%


Purity: 98% CAS No.: 79669-50-4 Molecular Formula: C9H9BrO2 Formula Weight: 229.07 Physical Form: Powder Appearance: Pale yellow MDL No.: MFCD09839555

Formula: C₉H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD09839555
CAS Number: 79669-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR