Methyl-2-bromo-3-methylbenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-bromo-3-methylbenzoate

H61831-14 H61831-22
CAAAH61831-14EA 192.76 CAD
CAAAH61831-14 CAAAH61831-22
Methyl-2-bromo-3-methylbenzoate 98%
Methyl 2-bromo-3-methylbenzoate

Methyl 2-bromo-3-methylbenzoate, 98%


Purity: 98% CAS No.: 131001-86-0 Molecular Formula: C9H9BrO2 Formula Weight: 229.07 Physical Form: Liquid Appearance: Red Beilstein Reference: 4177165 MDL No.: MFCD00144762

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144762
CAS Number: 131001-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR