5-Chloro-1-methyl-4-nitroimidazole 95%

Supplier: Thermo Scientific Chemicals
Warning

H61824-03 H61824-06
CAAAH61824-03EA 59.88 CAD
CAAAH61824-03 CAAAH61824-06
5-Chloro-1-methyl-4-nitroimidazole 95%
5-Chloro-1-methyl-4-nitroimidazole

5-Chloro-1-methyl-4-nitroimidazole, 95%


Purity: 95% CAS No.: 4897-25-0 Molecular Formula: C4H4ClN3O2 Formula Weight: 161.55 Physical Form: Powder Appearance: White MDL No.: MFCD00233664

Formula: C₄H₄ClN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00233664
CAS Number: 4897-25-0
EINECS: 225-521-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR