4-(Methoxymethyl)benzeneboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Methoxymethyl)phenylboronic acid, (4-(Methoxymethyl)phenyl)boronic acid

H61795-06 H61795-14
CAAAH61795-06EA 70.31 CAD
CAAAH61795-06 CAAAH61795-14
4-(Methoxymethyl)benzeneboronic acid 97%
4-(Methoxymethyl)benzeneboronic acid

4-(Methoxymethyl)benzeneboronic acid, 97%


Purity: 97% CAS No.: 279262-11-2 Molecular Formula: C8H11BO3 Formula Weight: 165.99 Physical Form: Solid MDL No.: MFCD03788426

Formula: C₈H₁₁BO₃
MW: 165.983 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788426
CAS Number: 279262-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR