Ethyl-2-amino-5-methylthiophene-3-carboxylate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-5-methylthiophene-3-carboxylic acid ethyl ester, Ethyl 2-amino-5-methylthiophene-3-carboxylate

H61766-03 H61766-06
CAAAH61766-03EA 200.66 CAD
CAAAH61766-03 CAAAH61766-06
Ethyl-2-amino-5-methylthiophene-3-carboxylate 95%
Ethyl-2-amino-5-methylthiophene-3-carboxylate

Ethyl 2-amino-5-methylthiophene-3-carboxylate, 95%


Purity: 95% CAS No.: 4815-32-1 Molecular Formula: C8H11NO2S Formula Weight: 185.24 Physical Form: Solid MDL No.: MFCD01922142

Formula: C₈H₁₁NO₂S
MW: 185.241 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01922142
CAS Number: 4815-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR