Methyl-4-piperidinecarboxylate 98%

Supplier: Thermo Scientific Chemicals
Danger

H61754-14 H61754-22
CAAAH61754-14EA 93.22 CAD
CAAAH61754-14 CAAAH61754-22
Methyl-4-piperidinecarboxylate 98%
Methyl 4-piperidinecarboxylate

Methyl piperidine-4-carboxylate, 98%

Formula: C₇H₁₃NO₂
Boiling Pt: 85…90 °C
Density: 1.06
Flash Pt: 89 °C(192 °F)
Storage Temperature: Ambient
MDL Number: MFCD00190578
CAS Number: 2971-79-1
UN: 3267
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR