5-Nitro-2-picoline 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methyl-5-nitropyridine

H61750-06 H61750-14
CAAAH61750-06EA 88.16 CAD
CAAAH61750-06 CAAAH61750-14
5-Nitro-2-picoline 95%
5-Nitro-2-picoline

2-Methyl-5-nitropyridine, 95%

Formula: C₆H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04114179
CAS Number: 21203-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR