2-Amino-6-methyl-3-nitropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

H61729-06 H61729-14
CAAAH61729-06EA 60.2 CAD
CAAAH61729-06 CAAAH61729-14
2-Amino-6-methyl-3-nitropyridine 98%
2-Amino-6-methyl-3-nitropyridine

2-Amino-6-methyl-3-nitropyridine, 98%

Formula: C₆H₇N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00047443
CAS Number: 21901-29-1
UN: 2811
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR