1H-Pyrazolo[3,4-b]pyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61710-03
H61710-MD
CAAAH61710-03EA
203.82
CAD
CAAAH61710-03
CAAAH61710-MD
1H-Pyrazolo[3,4-b]pyridine 97%
1H-Pyrazolo[3,4-b]pyridine
1H-Pyrazolo[3,4-b]pyridine, 97%
Purity: 97%
CAS No.: 271-73-8
Molecular Formula: C6H5N3
Formula Weight: 119.12
Physical Form: Solid
MDL No.: MFCD05663981
Formula:
C6H5N3 Storage Temperature: Ambient |
MDL Number:
MFCD05663981 CAS Number: 271-73-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...