2-(2-Fluorophenyl)pyrrolidine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61699-03
H61699-MD
CAAAH61699-03EA
535.62
CAD
CAAAH61699-03
CAAAH61699-MD
2-(2-Fluorophenyl)pyrrolidine 95%
2-(2-Fluorophenyl)pyrrolidine
2-(2-Fluorophenyl)pyrrolidine, 95%
Purity: 95%
CAS No.: 72216-04-7
Molecular Formula: C10H12FN
Formula Weight: 165.21
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD02663474
Formula:
C₁₀H₁₂FN Storage Temperature: Refrigerator |
MDL Number:
MFCD02663474 CAS Number: 72216-04-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...