Methyl-2-fluoro-5-nitrobenzenecarboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-fluoro-5-nitrobenzenecarboxylate, Methyl 2-fluoro-5-nitrobenzoate

H61676-06 H61676-14
CAAAH61676-06EA 110.76 CAD
CAAAH61676-06 CAAAH61676-14 CAAAH61676-22
Methyl-2-fluoro-5-nitrobenzenecarboxylate 98%
Methyl-2-fluoro-5-nitrobenzenecarboxylate

Methyl 2-fluoro-5-nitrobenzoate, 98%


Purity: 98% CAS No.: 2965-22-2 Molecular Formula: C8H6FNO4 Formula Weight: 199.14 Physical Form: Solid MDL No.: MFCD06203689

Formula: C₈H₆FNO₄
MW: 199.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203689
CAS Number: 2965-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR