Methyl-2-fluoro-4-hydroxybenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-fluoro-4-hydroxybenzoate

H61675-03 H61675-06
CAAAH61675-03EA 268.6 CAD
CAAAH61675-03 CAAAH61675-06 CAAAH61675-MD
Methyl-2-fluoro-4-hydroxybenzoate 98%
Methyl-2-fluoro-4-hydroxybenzoate

Methyl 2-fluoro-4-hydroxybenzoate, 98%


Purity: 98% CAS No.: 197507-22-5 Molecular Formula: C8H7FO3 Formula Weight: 170.14 Physical Form: Solid MDL No.: MFCD09038282

Formula: C8H7FO3
MW: 170.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09038282
CAS Number: 197507-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR