6-Fluoroindole-3-carboxaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61665-03
H61665-06
CAAAH61665-03EA
79.63
CAD
CAAAH61665-03
CAAAH61665-06
6-Fluoroindole-3-carboxaldehyde 98%
6-Fluoroindole-3-carboxaldehyde
6-Fluoroindole-3-carboxaldehyde, 98%
Purity: 98%
CAS No.: 2795-41-7
Molecular Formula: C9H6FNO
Formula Weight: 163.15
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline solid
Appearance: Yellow
MDL No.: MFCD00069703
Formula:
C₉H₆FNO Storage Temperature: Refrigerator |
MDL Number:
MFCD00069703 CAS Number: 2795-41-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...