5-Chloromethyl-1H-tetrazole 95%

Supplier: Thermo Scientific Chemicals
Warning

H61643-03 H61643-06 H61643-MD
CAAAH61643-03EA 138.57 CAD
CAAAH61643-03 CAAAH61643-06 CAAAH61643-MD
5-Chloromethyl-1H-tetrazole 95%
5-Chloromethyl-1H-tetrazole

5-Chloromethyl-1H-tetrazole, 95%


Purity: 95% CAS No.: 55408-11-2 Molecular Formula: C2H3ClN4 Formula Weight: 118.53 Physical Form: Solid Appearance: White MDL No.: MFCD00512845

Formula: C2H3ClN4
MW: 118.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00512845
CAS Number: 55408-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR