Methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropionate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61641-03
H61641-06
CAAAH61641-03EA
254.38
CAD
CAAAH61641-03
CAAAH61641-06
CAAAH61641-MD
Methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropionate 98%
Methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropionate
Methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropionate, 98%
Purity: 98%
CAS No.: 32981-85-4
Molecular Formula: C17H17NO4
Formula Weight: 299.32
Physical Form: Crystalline powder
Appearance: White
Melting Point: 178-183°
Optical Rotation: -44° (c=1 in methanol)
MDL No.: MFCD00673331
Formula:
C₁₇H₁₇NO₄ Melting Pt: 178…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD00673331 CAS Number: 32981-85-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...