Ethyl-3-(4-bromophenyl)propanoate 95%

Supplier: Thermo Scientific Chemicals
Warning

H61633-06 H61633-14
CAAAH61633-06EA 176.96 CAD
CAAAH61633-06 CAAAH61633-14
Ethyl-3-(4-bromophenyl)propanoate 95%
Ethyl-3-(4-bromophenyl)propanoate

Ethyl 3-(4-bromophenyl)propionate, 95%


Purity: 95% CAS No.: 40640-98-0 Molecular Formula: C11H13BrO2 Formula Weight: 257.12 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD07772961

Formula: C₁₁H₁₃BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD07772961
CAS Number: 40640-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR